Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 60 Well HLA Terasaki Plates

Catalog No. 07001009
Click to view available options
Well Count:
60

Suitable for all applications for serological determination of HLA antigens

  • Suitable for all applications for serological determination of HLA antigens
  • Manufactured from crystal clear polystyrene
  • Stackable
  • Improved cell adhesion through physical surface treatment
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 10 L (Well)
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Tissue Culture
Type Microtest Plate
Well Count 60
For Use With (Application) HLA Testing
Includes Lid
No. per Case 150 pcs/foldable carton
Length (Metric) 83.2 mm
Width (Metric) 57.8 mm
Height (Metric) 8 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.