Description
Conductivity Probes and Multiparameter Cables. Order other probes separately.
- The 60510 ISE (pH, ORP, pH/ORP, ammonium, nitrate, chloride) cables ONLY work with the YSI Pro Plus handheld Any of the ISE sensors can be connected to this cable at any time
Specifications
Specifications
| Includes | Cable management kit |
| Length (English) | 787.4 in. |
| Length (Metric) | 20 m |