Learn More
YSI™ 60520 Pro Series DO Cable
Catalog No. 1339889 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Length (English) Cable:
3.2 ft.
13 ft.
328 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Cable
13-398-89 3.2 ft.
15-177-470 13 ft.
15-176-217 328 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-398-89 Supplier YSI™ Supplier No. 605201
Add to Cart
Add to Cart
Compatible with the YSI Pro Plus or Pro20 handhelds
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty.Measure pH quickly and easily with the 1001 sensor.The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time information.
Cables over 1-meter also come with the 603062 cable management kit.
Order Info
Cables do NOT come with the sensor
TRUSTED_SUSTAINABILITY
Specifications
Type Cable Assembly
For Use With (Application) Measure temperature and DO
Length (English) Cable 3.2 ft.
Length (Metric) Cable 1 m
Warranty 1 Year