Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
60S ribosomal protein L23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312980
This item is not returnable.
View return policy
Description
60S ribosomal protein L23 Polyclonal antibody specifically detects 60S ribosomal protein L23 in Mouse samples. It is validated for Western BlotSpecifications
60S ribosomal protein L23 | |
Polyclonal | |
PBS, 1% BSA, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
9349 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
MGC111167, MGC117346, MGC72008,60S ribosomal protein L17,60S ribosomal protein L23, ribosomal protein L17, ribosomal protein L23, rpL17 | |
Recombinant protein encompassing a sequence within the center region of human 60S ribosomal protein L23. The exact sequence is proprietary. | |
100 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction