Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

60S Ribosomal Protein L26 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE143825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1438-25UL 25 μL
ABE1438MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE1438-25UL Supplier MilliporeSigma Supplier No. ABE143825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-60S Ribosomal Protein L26 Antibody is a highly specific rabbit polyclonal antibody that targets 60S ribosomal protein L26 and has been tested in Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 60S Ribosomal Protein L26
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols RPL26
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to the first 95 amino acids from human 60S ribosomal protein L26.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects 60S ribosomal protein L26 in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.