Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 60S RPL5, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1439 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1439 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1439 Supplier MilliporeSigma™ Supplier No. ABE1439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

60S ribosomal protein L5 (UniProt P46777; also known as RPL5) is encoded by the RPL5 (also known as DBA6, MSTP030) gene (Gene ID 6125) in human. Ribosomal proteins (RPs) are normally involved in the assembly of the translational machinery-ribosomes essential for protein production. RPL5, RPL11 and the 5S rRNA comprise the 5S ribonucleoprotein particle (RNP), an essential subcomplex of the large ribosomal subunit. RPL5 binds the 5S rRNA, the 5S rRNA/RPL5 complex then localizes to the nucleolus, where it binds RPL11 and is integrated into the ribosome. In response to ribosomal stress (RS) or nucleolar stress, the L5/L11/5S pre-ribosomal complex is redirected to inhibit MDM2 E3 ligase activity, thereby promoting p53 stabilization and p53-dependent cell cycle arrest. RPL5 and RPL11 can also bind TAp73 N-terminal TA domain in an RS-independent manner and thereby prevent MDM2 association with TAp73 target gene promoters, resulting in enhanced TAp73 transcriptional activity and TAp73-dependent apoptosis induction and cell cycle arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 60S RPL5
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RPL5, DBA6, MSTP030
Gene Accession No. P46777
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an internal sequence within the C-terminal half of human 60S RPL5.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000960
Test Specificity This rabbit polyclonal antibody targets a highly conserved sequence within the C-terminal half of 60S RPL5 from multiple species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.