Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 666-15
GREENER_CHOICE

Catalog No. 566110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-611-0 10 mg
56-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-611-0 Supplier Tocris Bioscience™ Supplier No. 5661/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CREB inhibitor

666-15 is a potent and selective CREB inhibitor (IC50 = 81 nM). Inhibits cancer cell growth, without harming normal cells (up to 1 μM). Completely suppresses tumor growth in a breast cancer xenograft model.

Specifications

Chemical Name or Material 3-(3-Aminopropoxy)-N-[2-[[3-[[(4-chloro-2-hydroxyphenyl)amino]carbonyl]-2-naphthalenyl]oxy]ethyl]-2-naphthalenecarboxamide hydrochloride
CAS 1433286-70-4
Quantity 10 mg
Target Additional Transcription Factor Compounds
Molecular Formula C33H30ClN3O5.HCl
Formula Weight 620.52
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.