Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

67kDa Laminin Receptor Monoclonal Antibody (OTI1G3), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF811423

Encompass
Encompass_Preferred

Catalog No. 50-254-3948


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

67kDa Laminin Receptor Monoclonal antibody specifically detects 67kDa Laminin Receptor in Canine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

67kDa Laminin Receptor
Monoclonal
See Label
PBS with 8% trehalose and no preservative; pH 7.3
B0K035, Q6P444
Mouse
Protein A/G
RUO
100911069, 113115, 483997
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1G3
Unconjugated
MTFR2
37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37 kDa laminin receptor precursor {ECO:0000255; 37 kDa oncofetal antigen; 37/67 kDa laminin receptor; 37/67 kDa laminin receptor {ECO:0000255; 37LRP; 37LRP {ECO:0000255; 40S ribosomal protein SA; 40S ribosomal protein SA {ECO:0000255; 67 kDa laminin receptor; 67 kDa laminin receptor {ECO:0000255; 67kDa; 67LR; 67LR {ECO:0000255; AL022858; colon carcinoma laminin-binding protein; HAMAP-Rule:MF_03016}; ICAS; LAMBR; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); laminin receptor 1 (ribosomal protein SA); laminin receptor 1 {ECO:0000255; laminin-binding protein precursor p40; laminin-binding protein precursor p40 {ECO:0000255; LamR; lamR {ECO:0000255; LAMR1; Lamrl1; LBP; LBP/p40; LBP/p40 {ECO:0000255; LRP; LRP/LR; LRP/LR {ECO:0000255; MLR; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; OFA/iLRP; p40; P40-3; P40-3, functional; P40-8; P40-8, functional; ribosomal protein SA; Rpsa; rssa; SA; Small ribosomal subunit protein uS2
Full length human recombinant protein of human FAM54A produced in E.coli.
100 μg
Primary
Canine, Human, Rat
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.