Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HydroSpec™ Solvent K, Ricca Chemical
SDP

Catalog No. p-6980042 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Solvent KT
CAS Min % 5
CAS Max % 10, 30
CAS 7446-09-5, 67-66-3, 107-98-2
Physical Form Liquid
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN3092
Specific Gravity 1.12
Grade Karl Fischer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.