Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 6x-His Tag Monoclonal Antibody (4E3D10H2/E3), Alexa Fluor™ 647
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1135A647
Description
6x-His Tag Monoclonal Antibody for Western Blot, ICC/IF
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
Specifications
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
4E3D10H2/E3 | |
Alexa Fluor 647 | |
6His tag; 6X His; His Tag; Histidine Tag | |
Mixture of N-terminal and C-terminal 6x-His peptides, CG-HHHHHH and HHHHHH-GC. | |
50 μL | |
Primary | |
4°C, do not freeze | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction