Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 6x-His Tag Monoclonal Antibody (HIS.H8), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501122713
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2713 100 μg
50-112-2712 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2713 Supplier Invitrogen™ Supplier No. 14665782
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This HIS.H8 monoclonal antibody reacts with the 6X polyhistidine (6X His) tag. The H-H-H-H-H-H motif is frequently fused to the N- or C-terminus of proteins to facilitate purification (e.g., nickel affinity chromatography and immunoprecipitation) and/or detection. This antibody recognizes the 6X His tag anywhere within the protein of interest (i.e., N-terminus, C-terminus, or internal). Applications Reported: This HIS.H8 antibody has been reported for use in immunoprecipitation and western blotting. Applications Tested: This HIS.H8 antibody has been tested by immunoblotting of reduced cell lysate expressing a His-tagged protein at less than or equal to 1 μg/ mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 6x-His Tag
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HIS.H8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Tag
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.