Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6X His Tag Mouse anti-Tag, Biotin, Clone: 33D10.D2.G8, Rockland™
SDP

Catalog No. 501990701
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0701 Supplier Rockland Immunochemicals Supplier No. 200306382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against the 6X His motif and is useful in determining its presence in various assays. This monoclonal anti-6X His tag antibody detects over-expressed proteins containing the 6X His epitope tag. To date, this antibody has reacted with all His tagged proteins so far tested. In western blotting of bacterial extracts, the antibody does not cross-react with endogenous proteins. The antibody recognizes the His-tag (His-His-His-His-His-His) fused to either the amino- or carboxy-termini of targeted proteins in transfected or transformed cells.

The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.

Specifications

Antigen 6X His Tag
Applications ELISA, Fluorescence Resonance Energy Transfer (FRET), Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 33D10.D2.G8
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Mouse
Immunogen This antibody was produced in mice by repeated immunizations with 6X His epitope tag peptide H-H-H-H-H-H conjugated to KLH using maleimide.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.