Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 6x His Tag Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141168 Supplier Invitrogen™ Supplier No. PA5141168
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

6x His Tag Polyclonal Antibody for Western Blot, IP

The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 6x His Tag
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Rabbit
Immunogen Synthetic peptide immune sequence is HHHHHHC.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.