Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone 98.0+%, TCI America™
SDP

Catalog No. p-7122880
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 129722-34-5
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD06658540
InChI Key URHLNHVYMNBPEO-UHFFFAOYSA-N
Synonym 7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline
PubChem CID 10542064
IUPAC Name 7-(4-bromobutoxy)-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)OCCCCBr

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.