Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Amino-1,3-naphthalenedisulfonic acid, Tech.

Catalog No. AC163960025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163960025 2.5 kg Plastic Bottle
AC163961000 100 g Glass bottle
AC163965000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163960025 Supplier Thermo Scientific Chemicals Supplier No. 163960025
Add to Cart

CAS: 86-65-7 | C10H9NO6S2 | 303.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-65-7
Molecular Formula C10H9NO6S2
Molecular Weight (g/mol) 303.29
MDL Number MFCD00003991
InChI Key CMOLPZZVECHXKN-UHFFFAOYSA-N
Synonym amido-g-acid, 7-amino-1,3-naphthalenedisulfonic acid, amino-g-acid, 1,3-naphthalenedisulfonic acid, 7-amino, amino-g acid, 2-amino-6,8-disulfonaphthalene, unii-7c482fx29k, 2-aminonaphthalene-6,8-disulfonic acid, 2-naphthylamine-6,8-disulfonic acid, beta-naphthylamine-6,8-disulfonic acid
PubChem CID 6851
IUPAC Name 7-aminonaphthalene-1,3-disulfonic acid
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N

Specifications

Melting Point >300°C
Color Brown to White
Infrared Spectrum Authentic
Packaging Plastic Bottle
Quantity 2.5 kg
Beilstein 14, 784
Merck Index 15, 395
Solubility Information Solubility in water: soluble
Formula Weight 303.29
Percent Purity 60% (as is basis) Typically
Grade Technical
Physical Form Needles or Powder
Chemical Name or Material 7-Amino-1, 3-naphthalenedisulfonic acid, tech.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-689-2
RTECSNumber QJ6132000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.