Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Amino-2-benzoxazolinone 95.0+%, TCI America™
SDP

Catalog No. A28061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28061G 1 g
A2806250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28061G Supplier TCI America Supplier No. A28061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81282-60-2
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.137
MDL Number MFCD09753616
InChI Key CLCPWTXGFUIRJE-UHFFFAOYSA-N
Synonym 7-Amino-2-benzoxazolone, 7-Amino-2-hydroxybenzoxazole
PubChem CID 11147814
IUPAC Name 7-amino-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C(=C1)N)OC(=O)N2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 150.14
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-2-benzoxazolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.