Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminocephalosporanic Acid 97.0+%, TCI America™
SDP

Catalog No. A126625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A126625G 25 g
A12665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A126625G Supplier TCI America Supplier No. A126625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 957-68-6
Molecular Formula C10H12N2O5S
Molecular Weight (g/mol) 272.275
MDL Number MFCD00005177
InChI Key HSHGZXNAXBPPDL-HZGVNTEJSA-N
Synonym 7-aminocephalosporanic acid, 7-aca, 7-aminocephalosporinic acid, 7beta-aminocephalosporanic acid, 7-acs, unii-9xi67897rg, 7r-7-aminocephalosporanic acid, 7r-7-aminocephalosporanate, cephalosporanic acid, 7-amino, 6r,7r-3-acetoxymethyl-7-amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
PubChem CID 441328
ChEBI CHEBI:2255
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

Quantity 25 g
Formula Weight 272.28
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminocephalosporanic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.