Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminonitrazepam Solution, 1.0 mg/mL in Acetonitrile, Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. A9131ML Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mL

Catalog No. A9131ML

Supplier: MilliporeSigma A9131ML

Only null left
Add to Cart
Add to Cart
  • 7-Aminonitrazepam is a major urinary metabolite of nitrazepam, a benzodiazepine used to treat insomnia and sold under the trade names Mogadon and Alodorm.
  • This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug monitoring of nitrazepam use.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Aminonitrazepam
Packaging Ampule
CAS 2-3-2028
Molecular Formula C15H13N3O
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Clinical Testing
Molecular Weight (g/mol) 251.28
Quantity 1 mL
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.