Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromo-1-heptene, 97%

Catalog No. aah6412722
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 4117-09-3 | C7H13Br | 177.09 g/mol

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4117-09-3
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD01631137
InChI Key GNYDYUQVALBGGZ-UHFFFAOYSA-N
IUPAC Name 7-bromohept-1-ene

Specifications

Color Colorless to Yellow
Density 1.162 g/mL
Boiling Point 27°C (1 mmHg)
Flash Point 58°C (136°F)
Linear Formula Br(CH2)5CH=CH2
Refractive Index 1.466
Quantity 100 g
UN Number UN1993
Beilstein 1740861
Synonym 2-amino-1,3,5-triazine, amino-1,3,5-triazine, 2-amino-s-triazine, 1,3,5-triazine-2-ylamine, chembl89436, amino-s-triazine, s-triazine-2-amine, pubchem14498, 1,3,5 triazin-2-ylamine
Solubility Information Slightly soluble in water.
SMILES BrCCCCCC=C
PubChem CID 20077
Formula Weight 177.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Bromo-1-heptene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.