Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromobenzo[b]thiophene 98.0+%, TCI America™
SDP

Catalog No. B48291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48291G 1 g
B4829200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48291G Supplier TCI America Supplier No. B48291G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1423-61-6
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.09
MDL Number MFCD06657729
InChI Key NOICDPBEDNMHQK-UHFFFAOYSA-N
PubChem CID 12045538
IUPAC Name 7-bromo-1-benzothiophene
SMILES BrC1=C2SC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 109°C
Quantity 1 g
Synonym 7-bromobenzo b thiophene, 7-bromobenzothiophene, 7-bromo-benzo b thiophene, benzo b thiophene, 7-bromo, zlchem 1192, 7-bromobenzo-b thiophene, benzo b thiophene,7-bromo
Formula Weight 213.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Bromobenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.