Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromoheptanoic acid, 97%

Catalog No. AC453260010
Click to view available options
Quantity:
1 g

CAS: 30515-28-7 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30515-28-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
InChI Key JLPQXFFMVVPIRW-UHFFFAOYSA-N
Synonym 7-bromo-heptanoic acid, heptanoic acid, 7-bromo, 7-bromoenanthic acid, omega-bromoheptanoic acid, 7-bromoheptanoicacid, 5-bromopentylacetate;, 7-bromo heptanoic acid, acmc-1aezo, ksc226m8n
PubChem CID 121723
IUPAC Name 7-bromoheptanoic acid
SMILES C(CCCBr)CCC(=O)O

Specifications

Melting Point 29.0°C to 32.0°C
Boiling Point 121.0°C to 125.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material 7-Bromoheptanoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 250-225-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.