Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-6-nitro-4-hydroxyquinazoline 98.0+%, TCI America™
SDP

Catalog No. C233225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C233225G 25 g
C23325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C233225G Supplier TCI America Supplier No. C233225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53449-14-2
Molecular Formula C8H4ClN3O3
Molecular Weight (g/mol) 225.588
MDL Number MFCD07772904
InChI Key URDYTQYZXZKBQT-UHFFFAOYSA-N
Synonym 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
PubChem CID 12441237
IUPAC Name 7-chloro-6-nitro-1H-quinazolin-4-one
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O

Specifications

Melting Point 303°C
Quantity 25 g
Formula Weight 225.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-6-nitro-4-hydroxyquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.