Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloroquinaldine 98.0+%, TCI America™
SDP

Catalog No. C145325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C145325G 25 g
C14535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C145325G Supplier TCI America Supplier No. C145325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4965-33-7
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.63
MDL Number MFCD00075436
InChI Key WQZQFYRSYLXBGP-UHFFFAOYSA-N
Synonym 7-Chloro-2-methylquinoline
PubChem CID 138395
IUPAC Name 7-chloro-2-methylquinoline
SMILES CC1=CC=C2C=CC(Cl)=CC2=N1

Specifications

Melting Point 76°C
Boiling Point 87°C
Quantity 25 g
Formula Weight 177.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloroquinaldine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.