Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloroquinoline 98.0+%, TCI America™
SDP

Catalog No. C308525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 612-61-3
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00956370
InChI Key QNGUPQRODVPRDC-UHFFFAOYSA-N
Synonym quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
PubChem CID 521963
IUPAC Name 7-chloroquinoline
SMILES C1=CC2=C(C=C(C=C2)Cl)N=C1

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 268°C
Quantity 25 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 7-Chloroquinoline
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.