Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Diethylamino-4-methylcoumarin, 99%

Click to view available options
Quantity:
100g
500g
91-44-1
C14H17NO2
231.295
MFCD00006864
AFYCEAFSNDLKSX-UHFFFAOYSA-N
7-diethylamino-4-methylcoumarin,coumarin 1,7-diethylamino-4-methyl-2h-chromen-2-one,coumarin 47,coumarin 460,4-methyl-7-diethylamino coumarin,blancophor aw,blancophor ffg,4-methyl-7-diethylaminocoumarin,hakkol p
7050
CHEBI:51938
7-(diethylamino)-4-methylchromen-2-one
CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C
91-44-1
C14H17NO2
231.295
MFCD00006864
AFYCEAFSNDLKSX-UHFFFAOYSA-N
7-diethylamino-4-methylcoumarin,coumarin 1,7-diethylamino-4-methyl-2h-chromen-2-one,coumarin 47,coumarin 460,4-methyl-7-diethylamino coumarin,blancophor aw,blancophor ffg,4-methyl-7-diethylaminocoumarin,hakkol p
7050
CHEBI:51938
7-(diethylamino)-4-methylchromen-2-one
CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C
Applications
7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.

Solubility
Soluble in water, aqueous acid solutions, resins, varnishes, vinyls, ether, acetone, ordinary organic solvents, and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 72°C to 75°C
Quantity 100g
Beilstein 193303
Formula Weight 231.3
Percent Purity 99%
Chemical Name or Material 7-Diethylamino-4-methylcoumarin

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GN6370000

TSCA : TSCA