Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, 99%, Thermo Scientific Chemicals

Catalog No. aab21767mf
Click to view available options
Quantity:
50 mg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84806-27-9
Molecular Formula C6H6FN3O4S
Molecular Weight (g/mol) 235.189
MDL Number MFCD02682109
InChI Key JXLHNMVSKXFWAO-UHFFFAOYSA-N
Synonym sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
PubChem CID 2737780
IUPAC Name azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
SMILES C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Specifications

Melting Point ∼285°C to 295°C (decomposition)
Quantity 50 mg
Sensitivity Light sensitive
Formula Weight 235.19
Percent Purity 99%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonic acid ammonium salt
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.