Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoroisatin 98.0+%, TCI America™
SDP

Catalog No. F055125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F055125G 25 g
F05515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F055125G Supplier TCI America Supplier No. F055125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 317-20-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
InChI Key HGBGVEOXPHGSOS-UHFFFAOYSA-N
Synonym 7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion
PubChem CID 586418
IUPAC Name 7-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)F)NC(=O)C2=O

Specifications

Melting Point 195°C
Quantity 25 g
Formula Weight 165.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Fluoroisatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.