Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Hydroxy-1-indanone, 97%

Click to view available options
Quantity:
250mg
1g
6968-35-0
C9H8O2
148.161
MFCD06659676
HFMZPBSZKCDKOR-UHFFFAOYSA-N
7-hydroxy-1-indanone,7-hydroxy-2,3-dihydro-1h-inden-1-one,7-hydroxyindan-1-one,1h-inden-1-one,2,3-dihydro-7-hydroxy,4h-inden-4-one, 1,2-dihydro-3-hydroxy,7-oxidanyl-2,3-dihydroinden-1-one,7-hydroxyindanone,acmc-20mvbk,pubchem23254,acmc-209wv8
248078
7-hydroxy-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC=C2O
6968-35-0
C9H8O2
148.161
MFCD06659676
HFMZPBSZKCDKOR-UHFFFAOYSA-N
7-hydroxy-1-indanone,7-hydroxy-2,3-dihydro-1h-inden-1-one,7-hydroxyindan-1-one,1h-inden-1-one,2,3-dihydro-7-hydroxy,4h-inden-4-one, 1,2-dihydro-3-hydroxy,7-oxidanyl-2,3-dihydroinden-1-one,7-hydroxyindanone,acmc-20mvbk,pubchem23254,acmc-209wv8
248078
7-hydroxy-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC=C2O
Quantity 250mg
Melting Point 109°C to 113°C
Beilstein 11085807
Formula Weight 148.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 7-Hydroxy-1-indanone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning