Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-2,2,5-trimethyl-2,3-dihydro-4H-chromen-4-one, 97%, Thermo Scientific™

Catalog No. RH01349CB
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 20052-60-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01313011
InChI Key RDRXIUHZBNIYAV-UHFFFAOYSA-N
Synonym 7-hydroxy-2,2,5-trimethyl-2,3-dihydro-4h-chromen-4-one, 7-hydroxy-2,2,5-trimethylchroman-4-one, 7-hydroxy-2,2,5-trimethyl-3h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2,2,5-trimethyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2,2,5-trimethyl, maybridge1_006242, 7-hydroxy-2,2,5-trimethyl-4-chromanone, 7-hydroxy-4-oxo-2,2,5-trimethylchroman, 2,3-dihydro-7-hydroxy-2,2,5-trimethyl-4h-chromen-4-one, 7-hydroxy-2,2,5-trimethyl-3,4-dihydro-2h-chromen-4-one
PubChem CID 2727964
IUPAC Name 7-hydroxy-2,2,5-trimethyl-3H-chromen-4-one
SMILES CC1=CC(=CC2=C1C(=O)CC(O2)(C)C)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Chemical Name or Material 7-Hydroxy-2,2,5-trimethyl-2,3-dihydro-4H-chromen-4-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.