Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxycoumarin, 99%

Catalog No. p-144592
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

CAS: 93-35-6 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-35-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006878
InChI Key ORHBXUUXSCNDEV-UHFFFAOYSA-N
Synonym 7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
PubChem CID 5281426
ChEBI CHEBI:27510
SMILES OC1=CC=C2C=CC(=O)OC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 228°C to 234°C
Color Beige-Brown to White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 18, V,1, 386
Merck Index 15, 10032
Solubility Information Solubility in water: 1g/100mL (100 c). Other solubilities: freely soluble in acetic acid and dilute alkalies,freely soluble in alcohol and chloroform,sparingly soluble in ether
IUPAC Name 7-hydroxychromen-2-one
Formula Weight 162.14
Percent Purity 99%
Physical Form Powder and Agglomerate
Chemical Name or Material 7-Hydroxycoumarin, 99%
Show More Show Less
EINECSNumber 202-240-3
RTECSNumber GN6820000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.