Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxyflavanone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
6515-36-2
C15H12O3
240.26
MFCD00017487
SWAJPHCXKPCPQZ-UHFFFAOYSA-N
7-hydroxyflavanone,7-hydroxy-2-phenylchroman-4-one,chembl97542,7-hydroxy-2-phenyl-4-chromanone,7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one,2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl,7-hydroxy-flavanone,d05zpo
1890
CHEBI:34483
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3
6515-36-2
C15H12O3
240.26
MFCD00017487
SWAJPHCXKPCPQZ-UHFFFAOYSA-N
7-hydroxyflavanone,7-hydroxy-2-phenylchroman-4-one,chembl97542,7-hydroxy-2-phenyl-4-chromanone,7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one,2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl,7-hydroxy-flavanone,d05zpo
1890
CHEBI:34483
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3
Quantity 1g
Packaging Glass bottle
Color Yellow
Melting Point 189°C to 197°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 18, V,2, 189
Formula Weight 240.26
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 7-Hydroxyflavanone, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.