Learn More
7-Ketocholesterol Antibody (3F7), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259357F
Catalog No. NBP259357F
Only null left
Add to Cart
Description
Description
7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
7-Ketocholesterol
Monoclonal
FITC
PBS with 0.05% Sodium Azide
Mouse
Protein G purified
Primary
Non-species specific
Purified
ELISA, Immunocytochemistry, Immunofluorescence
3F7
ELISA, Immunocytochemistry/Immunofluorescence
7KC, 7-KC, 7KC-Modified KLH
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for 7-ketocholesterol modified proteins.
Store at 4C in the dark.
IgG3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.