Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 7-Ketocholesterol Monoclonal Antibody (7E1), APC
GREENER_CHOICE

Catalog No. PIMA545793
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545793 Supplier Invitrogen™ Supplier No. MA545793
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45793 was sufficient for detection of 7-Ketocholesterol in 2 μg of 7-ketocholesterol conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for 7-ketocholesterol modified proteins.

7-Ketocholesterol is a major oxidation product of cholesterol found in human atherosclerotic plaque and is more atherogenic than cholesterol in some animal studies. 7-Ketocholesterol can inhibit cholesterol 7alpha-hydroxylase, the rate-limiting step in bile acid biosynthesis, as well as strongly inhibiting HMG-CoA reductase, the rate-limiting enzyme in cholesterol biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 7-Ketocholesterol
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7E1
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Alias 7KC; 7-KC; 7KC-Modified KLH
Host Species Mouse
Immunogen Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.