Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals

Catalog No. AAH6618806
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1810-74-8
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD00627122
InChI Key VNIQAUZZZWOJPT-UHFFFAOYSA-N
Synonym 7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 2,2,4-trimethyl-7-methoxy-1,2-dihydroquinoline, quinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethyl, 1,2-dihydro-2,2,4-trimethyl-7-methoxyquinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethylquinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro-quinoline, quinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro
PubChem CID 643478
IUPAC Name 7-methoxy-2,2,4-trimethyl-1H-quinoline
SMILES CC1=CC(NC2=C1C=CC(=C2)OC)(C)C

Specifications

Quantity 5 g
Beilstein 1426616
Formula Weight 203.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.