Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxycoumarin-4-Acetic Acid, PROMO, Thermo Scientific Chemicals

Catalog No. J6247503PM
Click to view available options
Quantity:
1 g

Precursor to coumarin-type fluorescent reagents

Applications

7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

Solubility

Soluble in acetone and dimethyl sulfoxide.

Notes

Desiccate at -20°C. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62935-72-2
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.21 g/mol
MDL Number MFCD00009774
InChI Key ZEKAXIFHLIITGV-UHFFFAOYSA-N
Synonym 7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
PubChem CID 342221
ChEBI CHEBI:51666
IUPAC Name 2-(7-methoxy-2-oxochromen-4-yl)acetic acid
SMILES COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1

Specifications

Melting Point 193°C
Color Green
Flash Point 87°C (189°F)
Quantity 1 g
Solubility Information Soluble in acetone and dimethyl sulfoxide.
Physical Form Crystalline Solid
Chemical Name or Material 7-Methoxycoumarin-4-acetic acid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.