Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxycoumarin, 98+%

Catalog No. aaa1544106
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 531-59-9 | C10H8O3 | 176.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-59-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00006876
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
PubChem CID 10748
ChEBI CHEBI:5679
IUPAC Name 7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 141728
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity ≥98%
Chemical Name or Material 7-Methoxycoumarin
EINECSNumber 208-513-3
RTECSNumber DJ3100380
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.