Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methyladenine 98%, Thermo Scientific™

Catalog No. AC325861000
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC325861000 100 mg
AC325862500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325861000 Supplier Thermo Scientific Chemicals Supplier No. 325861000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-69-3
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.157
InChI Key HCGHYQLFMPXSDU-UHFFFAOYSA-N
Synonym 7-methyladenine, 7-methyl-7h-purin-6-amine, 6-amino-7-methylpurine, n7-methyladenine, adenine, 7-methyl, 7h-purin-6-amine, 7-methyl, 7h-purin-6-amine, 7-methyl-9ci, 7-methyl-7h-adenine, 7-methyl-6-purinamine
PubChem CID 71593
ChEBI CHEBI:28921
IUPAC Name 7-methylpurin-6-amine
SMILES CN1C=NC2=C1C(=NC=N2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Quantity 100 mg
Beilstein 26, V,17, 126
Formula Weight 149.17
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 7-Methyladenine, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.