Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methylquinoline, 97%

Catalog No. p-7052067
Click to view available options
Quantity:
0.25 g
1 g

CAS: 612-60-2 | C10H9N | 143.189 g/mol

7-methylquinoline has been bioassayed as tumor initiator on the skin of Sencar mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-60-2
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006805
InChI Key KDYVCOSVYOSHOL-UHFFFAOYSA-N
Synonym quinoline, 7-methyl, m-toluquinoline, unii-eow995k43c, 7-methyl quinoline, ccris 2895, quinolinol,7-methyl, 7-methyl-quinoline, 7-methylchinolin, 7-methylchinolin;, pubchem5811
PubChem CID 11927
IUPAC Name 7-methylquinoline
SMILES CC1=CC2=C(C=CC=N2)C=C1

Specifications

Density 1.07 g/mL
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Quantity 0.25 g
Beilstein 110317
Solubility Information Insoluble in water.
Formula Weight 143.19
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 7-Methylquinoline
Hazard Category H302-H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-316-2
RTECSNumber VC0560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.