Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methylxanthine, 98%

Catalog No. p-7030498
Click to view available options
Quantity:
50 mg
250 mg

CAS: 552-62-5 | C6H6N4O2 | 166.14 g/mol

It is used to metabolite theophylline and caffeine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to metabolite theophylline and caffeine.

Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-62-5
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00037979
InChI Key PFWLFWPASULGAN-UHFFFAOYSA-N
Synonym 7-methylxanthine, heteroxanthine, heteroxanthin, 7-methylxanthin, 7-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-7-methylpurine, xanthine, 7-methyl, 3,7-dihydro-7-methyl-1h-purine-2,6-dione, 7-methyl-3,7-dihydro-1h-purine-2,6-dione, 1h-purine-2,6-dione, 3,7-dihydro-7-methyl
PubChem CID 68374
ChEBI CHEBI:48991
SMILES CN1C=NC2=C1C(=O)NC(=O)N2

Specifications

Melting Point >300°C
Quantity 50 mg
Solubility Information Soluble in dimethyl sulfoxide. Insoluble in water.
IUPAC Name 7-methyl-3H-purine-2,6-dione
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 7-Methylxanthine
EINECSNumber 209-019-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.