Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Octyn-1-ol

Catalog No. SYX002051G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002051G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002051G Supplier Sigma Aldrich Supplier No. SYX002051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloro-3-iodopropane undergoes asymmetric α-alkylation with n-sulfinyl imidates to yield 2-substituted n-tert-butanesulfinyl-5-chloropentanimidates. electroreduction of 1-chloro-3-iodopropane at glassy carbon electrode in dimethylformamide containing tetra-n-butylammonium perchlorate has been investigated by cyclic voltammetry. it also participates in conjugate addition of alkyl iodides to α,α-unsaturated nitriles in water.1-chloro-3-iodopropane has been used in the synthesis of:
  • N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (gar tfase) and aminoimidazole carboxamide ribonucleotide transformylase (aicar tfase)
  • Interesting αproton spongeα type molecule quino[7,8-h]quinoline

Specifications

CAS 871-91-0
Molecular Formula C8H14O
Linear Formula C8H14O
MDL Number MFCD01632137
Quantity 100 g
Molecular Weight (g/mol) 126.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.