Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
7% Perchloric Acid in Centrifuge Tubes, Cenmed™

Click to view available options
Quantity:
50 Tubes
Description
- 7% concentration is carefully formulated to meet the demands of precise laboratory work, providing an ideal environment for various applications
- Packaged in centrifuge tubes, this solution enhances safety and convenience during experiments
- Stand out in your lab practices with a reliable and contained reagent, purposefully crafted to meet the specific requirements of precise laboratory applications
Specifications
Specifications
Physical Form | Liquid |
CAS | 67-63-0 |
Color | Clear |
Quantity | 50 Tubes |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction