Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Tetradecanol 96.0+%, TCI America™
SDP

Catalog No. T17595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17595G 5 g
T17591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17595G Supplier TCI America Supplier No. T17595G

Chemical Identifiers

CAS 3981-79-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.393
MDL Number MFCD00049210
InChI Key FFAQYGTZIMVBFJ-UHFFFAOYSA-N
PubChem CID 549954
IUPAC Name tetradecan-7-ol
SMILES CCCCCCCC(CCCCCC)O

Specifications

Melting Point 42°C
Color White
Boiling Point 156°C
Quantity 5 g
Formula Weight 214.39
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Tetradecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.