Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Tetradecyne 97.0+%, TCI America™
SDP

Catalog No. T076225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T076225ML 25 mL
T07625ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T076225ML Supplier TCI America Supplier No. T076225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35216-11-6
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00041662
InChI Key AFNWSIIBAYUTTL-UHFFFAOYSA-N
PubChem CID 141979
IUPAC Name tetradec-7-yne
SMILES CCCCCCC#CCCCCCC

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 mL
Formula Weight 194.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Tetradecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.