Learn More
SHOWA 707HVO Orange Nitrile Gloves
Catalog No. 19395318 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-395-318 8
19-395-316 6
19-395-317 7
19-395-319 9
19-395-320 10
19-395-321 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-395-318 Supplier SHOWA Supplier No. 707HVO08
This item is not returnable. View return policy
The 707HVO gloves are highly tactile and fit like a second skin, all whilst protecting against grease, chemicals, and abrasion.
  • Fluorescent
  • Lightweight
  • Chlorinated
  • Eco Best Technology (EBT)
  • Unlined
  • Unsupported
Specifications
Certifications/Compliance (Europe) EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016
Color Orange
Size 8
Glove Cuff Style Rolled
Model 707HVO