Description
- Fluorescent
- Lightweight
- Chlorinated
- Eco Best Technology (EBT)
- Unlined
- Unsupported
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016 |
| Color | Orange |
| Size | 10 |
| Glove Cuff Style | Rolled |
| Model | 707HVO |