Learn More
Description
- Designed for sensitive skin, the 7502PF nitrile disposable glove is powder, latex, and accelerator-free.
- The 2.5 mil nitrile provides good chemical protection from a wide array of chemical hazards.
- And featuring SHOWA's revolutionary Eco Best Technology (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
Specifications
Specifications
| Certifications/Compliance | ISO 374-1:2016, ISO 374-5:2016 |
| Accelerator Free | Yes |
| Applications | Laboratory |
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Latex Free | Yes |
| Material | Nitrile |
| Powdered | Powder Free |
| Size | Large |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.