Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7H-Benz[de]anthracen-7-one, 99%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
82-05-3
C17H10O
230.266
MFCD00003585
HUKPVYBUJRAUAG-UHFFFAOYSA-N
benzanthrone,7h-benz de anthracen-7-one,benzanthrenone,ms-benzanthrone,dye, benzanthrone,benzoanthrone,mesobenzanthrone,7h-benzo de anthracen-7-one,1,9-benzanthrone
6697
benzo[a]phenalen-7-one
C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O
82-05-3
C17H10O
230.266
MFCD00003585
HUKPVYBUJRAUAG-UHFFFAOYSA-N
benzanthrone,7h-benz de anthracen-7-one,benzanthrenone,ms-benzanthrone,dye, benzanthrone,benzoanthrone,mesobenzanthrone,7h-benzo de anthracen-7-one,1,9-benzanthrone
6697
benzo[a]phenalen-7-one
C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O
Melting Point 171°C to 175°C
Quantity 25g
Beilstein 1455646
Merck Index 14,1063
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 7H-Benz[de]anthracen-7-one

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CX5075000

TSCA : TSCA