Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 7rh
GREENER_CHOICESDP

Catalog No. 58605 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 58-605 Supplier Tocris Bioscience™ Supplier No. 5860/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective DDR1 inhibitor; orally bioavailable

7rh is a potent and selective DDR1 inhibitor (IC50 = 6.8 nM). Exhibits selectivity for DDR1 over DDR2, Bcr-Abl and c-Kit (IC50 values are 101.4 and 355 nM and > 10 μM, respectively) and a range of other kinases. Inhibits proliferation of cancer cell lines expressing high levels of DDR1 in vitro and suppresses cancer cell invasion, adhesion and tumorigenicity. Orally bioavailable.

Specifications

Chemical Name or Material 4-Ethyl-N-[3-[(4-methyl-1-piperazinyl)methyl]-5-(trifluoromethyl)phenyl]-3-(2-pyrazolo[1,5-a]pyrimidin-6-ylethynyl)benzamide
CAS 1429617-90-2
Quantity 5 mg
Molecular Formula C30H29F3N6O
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.