Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 7X™-PF-O-Matic Detergent
GREENER_CHOICE

Catalog No. p-8820009
Click to view available options
Volume (English):
1 gal.
4 x 1 gal.
55 gal.

7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

  • Environmentally safe and phosphate free
  • Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available
  • Stable for long-term storage
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
pH 7 to 8
Appearance Dark Yellow
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, automatic washers
Quantity 4 Pk.
Volume (English) 4 x 1 gal.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.