Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-(3-Chlorostyryl)caffeine, Tocris Bioscience™

Catalog No. p-4783696
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective A2A antagonist. Also MAO-B inhibitor

Selective adenosine A2A receptor antagonist and monoamine oxidase B (MAO-B) inhibitor (Ki values are 54 and 28200 nM at rat A2A and A1 receptors respectively and Ki ∽ 100 nM at MAO-B). Potently protects against quinolinic acid -induced (Cat. No. 0225) neuronal damage and is neuroprotective in the MPTP model of Parkinson's disease.

Chemical Identifiers

CAS 147700-11-6
Molecular Formula C32H30Cl2N8O4
Molecular Weight (g/mol) 661.544
InChI Key YMUNFKYGBSLPLQ-UHFFFAOYSA-N
Synonym 1h-purine-2,6-dione,8-1e-2-3-chlorophenyl ethenyl-3,7-dihydro-1,3,7-trimethyl, acmc-20n59d
PubChem CID 57348597
IUPAC Name 8-[2-(3-chlorophenyl)ethenyl]-1,3,7-trimethylpurine-2,6-dione
SMILES CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl.CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl

Specifications

Quantity 10 mg
Formula Weight 330.77
Percent Purity >99%
Chemical Name or Material 8-(3-Chlorostyryl)caffeine
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.